5-Bromo-6-fluoro-3-methyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

19791-500MG 19791-1G 19791-5G 19791-25G
77324-234EA 150.38 USD
77324-234 77345-380 77360-520 77383-994
5-Bromo-6-fluoro-3-methyl-1H-indazole ≥95%
5-Bromo-6-fluoro-3-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₆BrFN₂
MW: 229.049 g/mol
Storage Temperature: Ambient
CAS Number: 864773-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR