2-Chloro-6-(trifluoromethyl)benzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

19790-500MG 19790-1G 19790-5G 19790-25G
77324-232EA 111.24 USD
77324-232 77343-286 77360-518 77372-428
2-Chloro-6-(trifluoromethyl)benzoic acid methyl ester ≥97%
2-Chloro-6-(trifluoromethyl)benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD14698012
CAS Number: 1214386-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR