Ethyl 6-isopropoxy-5-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

19774-500MG 19774-1G 19774-25G 19774-5G
77324-228EA 298.7 USD
77324-228 77348-160 77383-992 77372-424
Ethyl 6-isopropoxy-5-methylnicotinate ≥97%
Ethyl 6-isopropoxy-5-methylnicotinate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD32706991
CAS Number: 1188396-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR