5-(Aminocarbonyl)-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19651-500MG 19651-1G 19651-5G 19651-25G
77324-224EA 201.88 USD
77324-224 77348-158 77360-514 77383-988
5-(Aminocarbonyl)-2-methoxyphenylboronic acid ≥97%
5-(Aminocarbonyl)-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₄
MW: 194.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056744
CAS Number: 1127647-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR