N-(3-Bromo-4-methoxyphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

19566-500MG 19566-1G 19566-5G 19566-25G
77324-222EA 244.11 USD
77324-222 77348-156 77360-512 77383-986
N-(3-Bromo-4-methoxyphenyl)acetamide ≥97%
N-(3-Bromo-4-methoxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04094340
CAS Number: 6943-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR