Borate(1-), (4-bromo-2,5-dimethylphenyl)trifluoro-, potassium (1:1), (T-4)- ≥95%

Supplier: AOBChem USA

Warning

19465-500MG 19465-1G 19465-5G 19465-25G
77324-216EA 97.85 USD
77324-216 77342-652 77360-506 77372-416
Borate(1-), (4-bromo-2,5-dimethylphenyl)trifluoro-, potassium (1:1), (T-4)- ≥95%
Borate(1-), (4-bromo-2,5-dimethylphenyl)trifluoro-, potassium (1:1), (T-4)-

Precaución: for Research Use Only

Formula: C₈H₈BBrF₃K
Storage Temperature: Ambient
MDL Number: MFCD32706989
CAS Number: 1189097-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR