1,2,4-Tribromo-5-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1,2,4-Tribromo-5-iodobenzene

193717-1G 193717-5G 193717-25G
77341-402EA 84.46 USD
77341-402 77360-496 77372-410
1,2,4-Tribromo-5-iodobenzene ≥95%
1,2,4-Tribromo-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₃I
MW: 440.70 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18400058
CAS Number: 366496-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR