4-(4-Bromo-2,6-difluorobenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

19332-500MG 19332-1G 19332-5G 19332-25G
77324-210EA 224.54 USD
77324-210 77348-150 77360-494 77383-978
4-(4-Bromo-2,6-difluorobenzyl)morpholine ≥97%
4-(4-Bromo-2,6-difluorobenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD18910068
CAS Number: 1092563-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR