4-Bromo-2,6-dimethoxyphenol ≥95%

Supplier: AOBChem USA

Warning

19302-500MG 19302-1G 19302-5G 19302-25G
77324-208EA 82.4 USD
77324-208 77342-138 77360-492 77372-408
4-Bromo-2,6-dimethoxyphenol ≥95%
4-Bromo-2,6-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD18483099
CAS Number: 70654-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR