1-Acetyl-5-bromoindazole ≥97%

Supplier: AOBChem USA

Warning

19292-500MG 19292-1G 19292-5G 19292-25G
77324-206EA 191.58 USD
77324-206 77348-148 77360-490 77383-976
1-Acetyl-5-bromoindazole ≥97%
1-Acetyl-5-bromoindazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD17167291
CAS Number: 152626-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR