1-(5,6-Dimethyl-3-pyridinyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

192311-500MG 192311-1G 192311-5G 192311-25G
77324-204EA 196.73 USD
77324-204 77348-146 77360-488 77383-974
1-(5,6-Dimethyl-3-pyridinyl)ethanone ≥95%
1-(5,6-Dimethyl-3-pyridinyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Ambient
CAS Number: 1211535-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR