2,2-Dibromo-1-(4-chlorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

192108-500MG 192108-1G 192108-5G 192108-25G
77324-202EA 173.04 USD
77324-202 77346-436 77360-486 77383-972
2,2-Dibromo-1-(4-chlorophenyl)ethanone ≥95%
2,2-Dibromo-1-(4-chlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅Br₂ClO
MW: 312.38 g/mol
Storage Temperature: Ambient
CAS Number: 13651-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR