(3-Bromo-4-fluorophenoxy)(tert-butyl)dimethylsilane ≥95%

Supplier: AOBChem USA

Warning

19129-500MG 19129-1G 19129-5G 19129-25G
77324-198EA 234.84 USD
77324-198 77348-142 77360-482 77383-968
(3-Bromo-4-fluorophenoxy)(tert-butyl)dimethylsilane ≥95%
(3-Bromo-4-fluorophenoxy)(tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₂H₁₈BrFOSi
MW: 305.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18910242
CAS Number: 1037088-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR