3,5-Dibromo-2-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

18877-500MG 18877-1G 18877-5G 18877-25G
77324-196EA 154.5 USD
77324-196 77345-530 77360-470 77383-966
3,5-Dibromo-2-ethoxypyridine ≥97%
3,5-Dibromo-2-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇Br₂NO
MW: 280.945 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663708
CAS Number: 856852-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR