2-Bromo-3-carboxypyridine-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

18831-500MG 18831-1G 18831-5G 18831-25G
77324-194EA 199.82 USD
77324-194 77348-140 77360-468 77383-964
2-Bromo-3-carboxypyridine-5-boronic acid ≥97%
2-Bromo-3-carboxypyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrNO₄
MW: 245.823 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181656
CAS Number: 1451393-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR