N-(4-Fluoro-3-(trifluoromethyl)phenyl)acetamide ≥95%

Supplier: AOBChem USA

Warning

18828-1G 18828-5G 18828-25G
77340-596EA 60.77 USD
77340-596 77360-466 77372-396
N-(4-Fluoro-3-(trifluoromethyl)phenyl)acetamide ≥95%
N-(4-Fluoro-3-(trifluoromethyl)phenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₇F₄NO
MW: 221.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11616084
CAS Number: 2145-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR