tert-Butyl 3-(4-bromophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

187748-500MG 187748-1G 187748-5G 187748-25G
77324-192EA 93.73 USD
77324-192 77342-500 77360-464 77372-394
tert-Butyl 3-(4-bromophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%
tert-Butyl 3-(4-bromophenyl)-3-hydroxyazetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₃
MW: 328.2016 g/mol
Storage Temperature: Ambient
CAS Number: 1357614-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR