tert-Butyl 3-hydroxy-3-phenylazetidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

187746-500MG 187746-1G 187746-5G 187746-25G
77324-190EA 101.97 USD
77324-190 77343-028 77360-462 77372-392
tert-Butyl 3-hydroxy-3-phenylazetidine-1-carboxylate ≥95%
tert-Butyl 3-hydroxy-3-phenylazetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MW: 249.3055 g/mol
Storage Temperature: Ambient
CAS Number: 398489-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR