5-Chloro-4-hydroxy-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18750-500MG 18750-1G 18750-5G
77338-982EA 540.75 USD
77338-982 77348-138 77372-388
5-Chloro-4-hydroxy-3-(trifluoromethyl)phenylboronic acid ≥97%
5-Chloro-4-hydroxy-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₃
MW: 240.372 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976972
CAS Number: 2121511-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR