2,4-Dichloro-5-methylacetanilide ≥97%

Supplier: AOBChem USA

Warning

18692-500MG 18692-1G 18692-5G 18692-25G
77324-188EA 141.11 USD
77324-188 77344-772 77360-458 77383-962
2,4-Dichloro-5-methylacetanilide ≥97%
2,4-Dichloro-5-methylacetanilide

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO
MW: 218.08 g/mol
Storage Temperature: Ambient
CAS Number: 57046-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR