1,5-Dichloro-2-ethyl-4-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

18685-500MG 18685-1G 18685-5G
77337-566EA 423.33 USD
77337-566 77348-136 77372-384
1,5-Dichloro-2-ethyl-4-nitrobenzene ≥97%
1,5-Dichloro-2-ethyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MW: 220.053 g/mol
Storage Temperature: Ambient
CAS Number: 49709-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR