3-Chloro-2,6-difluoropyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

18657-500MG 18657-1G 18657-5G
77336-772EA 394.49 USD
77336-772 77348-132 77372-380
3-Chloro-2,6-difluoropyridine-4-carboxylic acid ≥97%
3-Chloro-2,6-difluoropyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD20657450
CAS Number: 685517-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR