5-Bromo-6-methylpyridine-2-carboxylic acid isopropyl ester ≥97%

Supplier: AOBChem USA

Warning

18639-500MG 18639-1G 18639-5G 18639-25G
77324-180EA 199.82 USD
77324-180 77348-130 77360-452 77383-956
5-Bromo-6-methylpyridine-2-carboxylic acid isopropyl ester ≥97%
5-Bromo-6-methylpyridine-2-carboxylic acid isopropyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD19105359
CAS Number: 1122092-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR