Ethyl 5-Bromo-6-methylpyridine-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-6-methylpyridine-2-carboxylic acid ethyl ester

18638-500MG 18638-1G 18638-5G 18638-25G
77324-178EA 177.16 USD
77324-178 77346-576 77360-450 77383-954
Ethyl 5-Bromo-6-methylpyridine-2-carboxylate ≥97%
Ethyl 5-Bromo-6-methylpyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18382715
CAS Number: 1122090-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR