6-Bromo-3-chloro-2,4-difluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

18578-500MG 18578-1G 18578-5G 18578-25G
77324-174EA 106.09 USD
77324-174 77343-110 77360-444 77372-374
6-Bromo-3-chloro-2,4-difluorobenzoic acid ≥95%
6-Bromo-3-chloro-2,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂BrClF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11845974
CAS Number: 1160573-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR