2,2-Difluorobenzo[1,3]dioxole-4-boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2,2-Difluorobenzo[d][1,3]dioxol-4-yl)boronic acid, 2,2-Difluoro-benzo[1,3]dioxol-4-boronic acid

18511-500MG 18511-1G 18511-5G 18511-25G
77324-172EA 92.7 USD
77324-172 77342-456 77360-442 77372-370
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid ≥95%
2,2-Difluorobenzo[1,3]dioxole-4-boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₄
Storage Temperature: Ambient
MDL Number: MFCD08062343
CAS Number: 126120-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR