4-(Hydroxymethyl)-3-methylphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

18428-1G 18428-5G 18428-25G
77341-662EA 98.88 USD
77341-662 77360-436 77372-360
4-(Hydroxymethyl)-3-methylphenylboronic acid pinacol ester ≥95%
4-(Hydroxymethyl)-3-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18732622
CAS Number: 1160430-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR