2,5-Dibromo-4-pyridinylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

18354-500MG 18354-1G 18354-5G 18354-25G
77324-168EA 199.82 USD
77324-168 77348-126 77360-430 77383-950
2,5-Dibromo-4-pyridinylboronic acid pinacol ester ≥97%
2,5-Dibromo-4-pyridinylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BBr₂NO₂
MW: 362.853 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181637
CAS Number: 1451391-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR