2-(Benzyloxy)-5-bromo-3-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

18333-500MG 18333-1G 18333-5G
77324-166EA 314.15 USD
77324-166 77348-124 77372-354
2-(Benzyloxy)-5-bromo-3-fluorobenzoic acid ≥95%
2-(Benzyloxy)-5-bromo-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrFO₃
MW: 325.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28063070
CAS Number: 1003298-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR