5-Bromo-3-methyl-N-phenylpyridine-3-carboxamide ≥97%

Supplier: AOBChem USA

Warning

18323-500MG 18323-1G 18323-5G 18323-25G
77324-162EA 146.26 USD
77324-162 77345-148 77360-426 77383-948
5-Bromo-3-methyl-N-phenylpyridine-3-carboxamide ≥97%
5-Bromo-3-methyl-N-phenylpyridine-3-carboxamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD15475027
CAS Number: 213771-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR