2,4-Dichloro-6-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

18253-500MG 18253-1G 18253-5G 18253-25G
77324-158EA 258.53 USD
77324-158 77348-120 77360-422 77383-944
2,4-Dichloro-6-methoxybenzaldehyde ≥95%
2,4-Dichloro-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27928772
CAS Number: 78444-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR