3-Bromo-2,6-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

180365-1G 180365-5G 180365-25G
77340-980EA 65.92 USD
77340-980 77360-416 77372-348
3-Bromo-2,6-dimethylaniline ≥97%
3-Bromo-2,6-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD16658602
CAS Number: 53874-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR