2-Chloro-5-methoxy-pyridine-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-methoxyisonicotinaldehyde

17882-500MG 17882-1G 17882-5G
77324-152EA 350.2 USD
77324-152 77348-112 77372-340
2-Chloro-5-methoxy-pyridine-4-carbaldehyde ≥95%
2-Chloro-5-methoxy-pyridine-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD13189002
CAS Number: 1060801-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR