2-Ethoxy-3-methylpyridine-5-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

17870-500MG 17870-1G 17870-5G 17870-25G
77324-150EA 154.5 USD
77324-150 77345-528 77360-408 77383-938
2-Ethoxy-3-methylpyridine-5-carboxaldehyde ≥97%
2-Ethoxy-3-methylpyridine-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181614
CAS Number: 1224604-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR