2,4-Dibromo-6-trifluoromethylphenol ≥97%

Supplier: AOBChem USA

Warning

17754-500MG 17754-1G 17754-5G 17754-25G
77324-146EA 126.69 USD
77324-146 77344-102 77360-400 77372-334
2,4-Dibromo-6-trifluoromethylphenol ≥97%
2,4-Dibromo-6-trifluoromethylphenol

Precaución: for Research Use Only

Formula: C₇H₃Br₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD11226207
CAS Number: 392-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR