2,5-Dichloro-4-pyridinecarboxaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

17665-1G 17665-5G 17665-25G
77342-092EA 113.3 USD
77342-092 77360-396 77372-330
2,5-Dichloro-4-pyridinecarboxaldehyde oxime ≥97%
2,5-Dichloro-4-pyridinecarboxaldehyde oxime

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
MW: 191.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24450275
CAS Number: 102645-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR