5-Bromo-N-(diphenylmethylene)-2-pyrimidinamine ≥97%

Supplier: AOBChem USA

Warning

17512-500MG 17512-1G 17512-5G 17512-25G
77324-142EA 176.13 USD
77324-142 77346-502 77360-392 77383-932
5-Bromo-N-(diphenylmethylene)-2-pyrimidinamine ≥97%
5-Bromo-N-(diphenylmethylene)-2-pyrimidinamine

Precaución: for Research Use Only

Formula: C₁₇H₁₂BrN₃
MW: 338.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105354
CAS Number: 1072850-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR