N-(4-Bromo-2,3-dimethylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

17487-500MG 17487-1G 17487-25G 17487-5G
77324-140EA 294.58 USD
77324-140 77348-106 77383-930 77372-326
N-(4-Bromo-2,3-dimethylphenyl)acetamide ≥97%
N-(4-Bromo-2,3-dimethylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
MW: 242.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03931825
CAS Number: 22369-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR