3-Fluoro-4-(1-methylethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

17439-500MG 17439-1G 17439-5G 17439-25G
77324-136EA 87.55 USD
77324-136 77342-280 77360-386 77372-320
3-Fluoro-4-(1-methylethoxy)benzaldehyde ≥95%
3-Fluoro-4-(1-methylethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156459
CAS Number: 1021231-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR