2,3-Difluoro-4-formyl-benzyloxybenzene ≥97%

Supplier: AOBChem USA

Warning

17250-500MG 17250-1G 17250-5G 17250-25G
77324-134EA 113.3 USD
77324-134 77343-342 77360-382 77372-316
2,3-Difluoro-4-formyl-benzyloxybenzene ≥97%
2,3-Difluoro-4-formyl-benzyloxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13181654
CAS Number: 947279-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR