2-Bromo-3,6-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17204-500MG 17204-1G 17204-5G 17204-25G
77324-132EA 109.18 USD
77324-132 77343-232 77360-378 77372-312
2-Bromo-3,6-dichlorobenzaldehyde ≥97%
2-Bromo-3,6-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O
MW: 253.908 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520679
CAS Number: 1114809-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR