4-Tert-butyl-2-hydroxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

17129-500MG 17129-1G 17129-25G 17129-5G
77324-128EA 285.31 USD
77324-128 77348-100 77383-924 77372-310
4-Tert-butyl-2-hydroxyphenylboronic acid ≥95%
4-Tert-butyl-2-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414823
CAS Number: 1068155-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR