1-Bromo-5-fluoro-2-(phenylmethoxy)-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

17022-500MG 17022-1G 17022-5G 17022-25G
77324-124EA 161.71 USD
77324-124 77345-824 77360-372 77383-922
1-Bromo-5-fluoro-2-(phenylmethoxy)-3-(trifluoromethyl)benzene ≥97%
1-Bromo-5-fluoro-2-(phenylmethoxy)-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉BrF₄O
MW: 349.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755709
CAS Number: 130046-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR