3-Fluoro-N-methoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

17015-500MG 17015-1G 17015-5G 17015-25G
77324-122EA 204.97 USD
77324-122 77348-094 77360-368 77383-920
3-Fluoro-N-methoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%
3-Fluoro-N-methoxy-N-methyl-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₉F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD27949476
CAS Number: 329941-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR