1,3-Dibromo-2-butoxy-5-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

16994-1G 16994-5G 16994-25G
77340-590EA 60.77 USD
77340-590 77360-364 77372-300
1,3-Dibromo-2-butoxy-5-fluorobenzene ≥97%
1,3-Dibromo-2-butoxy-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD28976898
CAS Number: 2484888-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR