2-(5-Bromo-2-trifluoromethoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

16918-500MG 16918-1G 16918-5G 16918-25G
77324-114EA 144.2 USD
77324-114 77345-036 77360-356 77383-914
2-(5-Bromo-2-trifluoromethoxyphenyl)-1,3-dioxolane ≥95%
2-(5-Bromo-2-trifluoromethoxyphenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₃
MW: 313.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209337
CAS Number: 773093-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR