2,3-Dimethoxy-6-(phenylmethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16857-500MG 16857-1G 16857-5G
77339-164EA 556.2 USD
77339-164 77348-090 77372-292
2,3-Dimethoxy-6-(phenylmethoxy)phenylboronic acid ≥97%
2,3-Dimethoxy-6-(phenylmethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₇BO₅
MW: 288.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20923614
CAS Number: 786691-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR