1-(Benzyloxy)-3,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

16856-500MG 16856-1G 16856-5G 16856-25G
77324-110EA 267.8 USD
77324-110 77348-088 77360-352 77383-912
1-(Benzyloxy)-3,4-dimethoxybenzene ≥97%
1-(Benzyloxy)-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD12913409
CAS Number: 42138-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR