tert-Butyl(5-ethoxy-2-fluorophenoxy)dimethylsilane ≥97%

Supplier: AOBChem USA

Warning

16812-500MG 16812-1G 16812-5G 16812-25G
77324-108EA 166.86 USD
77324-108 77346-182 77360-350 77383-910
tert-Butyl(5-ethoxy-2-fluorophenoxy)dimethylsilane ≥97%
tert-Butyl(5-ethoxy-2-fluorophenoxy)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₄H₂₃FO₂Si
MW: 270.415 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976892
CAS Number: 577793-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR