2-(3-(Benzyloxy)-6-bromo-2-fluorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

16794-500MG 16794-1G 16794-5G 16794-25G
77324-106EA 144.2 USD
77324-106 77345-034 77360-348 77383-908
2-(3-(Benzyloxy)-6-bromo-2-fluorophenyl)-1,3-dioxolane ≥95%
2-(3-(Benzyloxy)-6-bromo-2-fluorophenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₆H₁₄BrFO₃
MW: 353.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631771
CAS Number: 2379321-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR